Nickel(II) phthalocyanine, Thermo Scientific Chemicals
Nickel(II) phthalocyanine, Thermo Scientific Chemicals
Nickel(II) phthalocyanine, Thermo Scientific Chemicals
Nickel(II) phthalocyanine, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel(II) phthalocyanine, Thermo Scientific Chemicals
CAS: 14055-02-8 | C32H16N8Ni | 571.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALF039453.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS107-21-1
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Infrared spectrumConforms
Sulfated ash=<0.005 %
Iron (Fe)=<0.5 ppm
Water=<0.1 % (K.F.)