Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF039369.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS51-44-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)203.5-209.5?C
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.5%