Manganese(II) phthalocyanine, Thermo Scientific Chemicals
Manganese(II) phthalocyanine, Thermo Scientific Chemicals
Manganese(II) phthalocyanine, Thermo Scientific Chemicals
Manganese(II) phthalocyanine, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Manganese(II) phthalocyanine, Thermo Scientific Chemicals
CAS: 14325-24-7 | C32H16MnN8 | 567.474 g/mol
Have Questions?
Catalog NumberQuantity
ALF039309.031 g
Catalog number ALF039309.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7449-74-3
IUPAC NameN-methyl-N-(trimethylsilyl)acetamide
Molecular FormulaC6H15NOSi
InChI KeyQHUOBLDKFGCVCG-UHFFFAOYSA-N
SMILESCN(C(C)=O)[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
Assay (GC)≥95.0%
FormLiquid
Refractive Index1.4370-1.4410 @ 20°C