Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ALF039214.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UV1 cm cell vs water
UVat 250 nm A: =<0.30
UVat 300 nm A: =<0.01
GC>=99.0 %
View more
It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.