Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number ALF039214.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS536-66-3
IUPAC Name4-(propan-2-yl)benzoic acid
Molecular FormulaC10H12O2
InChI KeyCKMXAIVXVKGGFM-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point116°C to 120°C
HPLC>=98.0 %
Appearance (Color)White
It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.