Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
CAS: 10060-13-6 | Cl4CuH12N2O2 | 277.454 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF039127.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS34732-09-7
IUPAC Name2,3,4-trichlorobenzene-1-sulfonyl chloride
Molecular FormulaC6H2Cl4O2S
InChI KeyJDAJYNHGBUXIKS-UHFFFAOYSA-N
SMILESClC1=CC=C(C(Cl)=C1Cl)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Melting Point (clear melt)57.0-66.0°C
FormPowder
Assay (GC)≥94.0%
Ammonium copper(II) chloride dihydrate is used in the determination iron of content in steel. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.