Iodine monochloride, Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ALF039104.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS32042-38-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.3040-1.3090 @ 20?C
ICl is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.