Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
Catalog number ALF038994.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS3694-45-9
IUPAC Name1-chloro-4-(isothiocyanatomethyl)benzene
Molecular FormulaC8H6ClNS
InChI KeyDEHXIHUIYSXZNH-UHFFFAOYSA-N
SMILESClC1=CC=C(CN=C=S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.6120-1.6170 @ 20°C