Petroleum ether 35/60, Thermo Scientific Chemicals
Petroleum ether 35/60, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether 35/60, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number ALF038985.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPetroleum ether 35/60
Name NoteHPLC Grade
CAS8032-32-4
Health Hazard 1H225-H304-H315-H319-H336
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
View more
Petroleum ether 35/60 is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.