Sulfuric acid, Environmental Grade, 93-98%, Thermo Scientific Chemicals
Sulfuric acid, Environmental Grade, 93-98%, Thermo Scientific Chemicals
Sulfuric acid, Environmental Grade, 93-98%, Thermo Scientific Chemicals
Sulfuric acid, Environmental Grade, 93-98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, Environmental Grade, 93-98%, Thermo Scientific Chemicals
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF038751.K42.5 L
Catalog number ALF038751.K4
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS7439-95-4
IUPAC Namemagnesium
Molecular FormulaMg
InChI KeyJLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Sulfuric acid has a strong dehydrating agent and is applied in many applications like the domestic acidic drain cleaner, electrolyte in lead-acid batteries and in various other cleansing agents. It is also useful for mineral processing, fertilizer manufacturing, petroleum purification, wastewater processing, and chemical synthesis. The catalytic decomposition of sulfuric acid when mixed with Cr/Fe oxides, shows a useful application in the sulfur iodine cycle for hydrogen production.