Magnesium phthalocyanine, tech. 90%, Thermo Scientific Chemicals
Magnesium phthalocyanine, tech. 90%, Thermo Scientific Chemicals
Magnesium phthalocyanine, tech. 90%, Thermo Scientific Chemicals
Magnesium phthalocyanine, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium phthalocyanine, tech. 90%, Thermo Scientific Chemicals
CAS: 1661-03-6 | C32H18MgN8O | 554.856 g/mol
Have Questions?
Catalog NumberQuantity
ALF038601.031 g
Catalog number ALF038601.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS20211-76-1
IUPAC Namelanthanum(3+) trichloride
Molecular FormulaCl3La
InChI KeyICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystals
Gravimetry64.5 to 70 %
Magnesium phthalocyanine is used in preparing membrane sensors for cyanide ions. Also used in preparation of membrane sensors for cyanide ions.