Boric acid, 99.99% (metals basis), Thermo Scientific Chemicals
Boric acid, 99.99% (metals basis), Thermo Scientific Chemicals
Boric acid, 99.99% (metals basis), Thermo Scientific Chemicals
Boric acid, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Catalog NumberQuantity
ALF036771.36500 g
Catalog number ALF036771.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentBa 0.002%
Trace Metals(as Pb) 5ppm
Impurity contentIO₃ 3ppm
pHof a 5% solution 6.0-9.2 at 25?
Assay (unspecified)99.0% KI min
View more
It is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is too employed in leather manufacture, electroplating, and cosmetics.