Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals
Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals
Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals
Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals
Used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits.
Have Questions?
Catalog NumberQuantity
ALF036737.22100 g
Catalog number ALF036737.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS5039-78-1
IUPAC Nametrimethyl({2-[(2-methylprop-2-enoyl)oxy]ethyl})azanium chloride
Molecular FormulaC9H18ClNO2
InChI KeyRRHXZLALVWBDKH-UHFFFAOYSA-M
SMILES[Cl-].CC(=C)C(=O)OCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (HPLC)≥67.0%
Assay (Titration ex Chloride)≥66.5%
FormLiquid
Refractive Index1.4610-1.4730 @ 20°C
Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.