Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
CAS: 584-84-9 | C9H6N2O2 | 174.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF036733.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS126-58-9
IUPAC Name2-{[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl}-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC10H22O7
InChI KeyTXBCBTDQIULDIA-UHFFFAOYSA-N
SMILESOCC(CO)(CO)COCC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point215°C to 225°C
Hydroxyl value1125 to 1325 mg KOH/g
Appearance (Form)Powder
Infrared spectrumConforms
View more
Toluene diisocyanate is utilized in polyurethane foams and tank trucks. It is also used in concrete sealers for aircraft, sealers and coatings in floor. It is employed as a crosslinking agent for nylon.