Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals
CAS: 584-84-9 | C9H6N2O2 | 174.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ALF036733.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialToluene-2,4-diisocyanate
CAS584-84-9
Health Hazard 1H315-H317-H319-H330-H334-H335-H351
Health Hazard 2GHS H Statement
H330-H334-H351-H315-H319-H317-H335-H204
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.
Health Hazard 3P201-P202-P260-P264b-P271-P272-P280-P281-P284-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P333+P313-P362-P501c
View more
Toluene diisocyanate is utilized in polyurethane foams and tank trucks. It is also used in concrete sealers for aircraft, sealers and coatings in floor. It is employed as a crosslinking agent for nylon.