Methyl cellulose, viscosity 4000 cPs
Methyl cellulose, viscosity 4000 cPs
Methyl cellulose, viscosity 4000 cPs
Methyl cellulose, viscosity 4000 cPs
Thermo Scientific Chemicals
Methyl cellulose, viscosity 4000 cPs
CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Catalog NumberQuantity
ALF036718.22100 g
Catalog number ALF036718.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl cellulose
Name NoteViscosity: 4000cPs
CAS9004-67-5
Recommended StorageAmbient temperatures
TSCAYes
View more
Methyl cellulose is used as a thickener and an emulsifier in various food and cosmetic products; as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. In paper and textile industries, it is used as a sizing, thereby protecting the fibers from water or oil.