Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Air Sensitive
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number ALF036694.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
AppearanceColorless to pale yellow liquid
Chemical Name or MaterialHydrobromic acid
ColorColorless to Yellow
CAS10035-10-6
Health Hazard 1H290-H314-H335
View more
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes, and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration, and dehydration, and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.