Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number ALF036664.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS6636-55-1
IUPAC Namemethyl 6-chloropyridine-2-carboxylate
Molecular FormulaC7H6ClNO2
InChI KeyTWUXBVMXSBEKHA-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.