Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%, Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number ALF036664.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS586-95-8
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder or fused solid
Assay (GC)≥98.5%
Water Content (Karl Fischer Titration)<0.5%
Melting Point (clear melt)52-60?C
Appearance (Color)White to cream or pale yellow
Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.