Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Reagent for organic synthesis
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number ALF036609.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThiourea
CAS62-56-6
Health Hazard 1H302-H350-H361d-H500
Health Hazard 2GHS H Statement
H351-H361-H302
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Health Hazard 3P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
View more
Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.