Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Reagent for organic synthesis
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number ALF036609.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS352303-67-4
IUPAC Name(2-fluoro-3-methoxyphenyl)boronic acid
Molecular FormulaC7H8BFO3
InChI KeyJCKZNMSBFBPDPM-UHFFFAOYSA-N
SMILESCOC1=CC=CC(B(O)O)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0%
Proton NMRConforms to structure
Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.