Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals
Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals
Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals
Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals
Zinc Granules; ACS; 99.8%; Alfa Aesar; -20 Mesh
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Catalog number ALF036602.A1
Price (MYR)
986.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
986.00
Add to cart
Specifications
Chemical Name or MaterialZinc
Name NoteFor analysis ACS
CAS7440-66-6
Recommended StorageAmbient temperatures
RTECS NumberZG8600000