Gallium(III) isopropoxide, mixture of oligomers, 99%, Thermo Scientific Chemicals
Gallium(III) isopropoxide, mixture of oligomers, 99%, Thermo Scientific Chemicals
Gallium(III) isopropoxide, mixture of oligomers, 99%, Thermo Scientific Chemicals
Gallium(III) isopropoxide, mixture of oligomers, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallium(III) isopropoxide, mixture of oligomers, 99%, Thermo Scientific Chemicals
CAS: 4452-61-3 | C3H8GaO | 129.819 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALF036583.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGallium(III) isopropoxide
Name NoteMixture of oligomers
CAS4452-61-3
Health Hazard 1H227-H315-H319
Health Hazard 2GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
View more
Gallium(III) isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.