Sucrose, ACS
Sucrose, ACS
Sucrose, ACS
Thermo Scientific Chemicals
Sucrose, ACS
CAS: 57-50-1 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF036508.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS51325-91-8
IUPAC Name2-{2-[(1E)-2-[4-(dimethylamino)phenyl]ethenyl]-6-methyl-4H-pyran-4-ylidene}propanedinitrile
Molecular FormulaC19H17N3O
InChI KeyYLYPIBBGWLKELC-RMKNXTFCSA-N
SMILESCN(C)C1=CC=C(C=CC2=CC(C=C(C)O2)=C(C#N)C#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to red-brown
Appearance (Form)Powder or crystals
UV Lambda max479 to 483 nm (in DMSO)
HPLC>=96.0 %
Molar absorption coefficient>=42000 l/(mol.cm) (at Lambda max)
View more
Sucrose is widely utilized in medications to carry a more pleasant taste to indigestible chemicals. It is used in many medical dosage forms such as chewable tablets, syrups, tablets, or glues. Sucrose is mainly used in foods nowadays. The fatty acid esters of sucrose show useful application in transdermal therapeutic systems. Sucrose-gradient centrifugation also plays an important role in the selective isolation of nocardia spp. from soil.