Sucrose, ACS
Sucrose, ACS
Sucrose, ACS
Thermo Scientific Chemicals
Sucrose, ACS
CAS: 57-50-1 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ALF036508.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS63231-69-6
IUPAC NameMolecular sieves 13X
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
FormPellets
Appearance (Color)White to pale yellow
CommentApparent density:≥37 lbs/ft³
CommentCO₂ capacity at 250 Torr: ≥17 lbs/100 lbs
Loss on ignition≤1.0% at 575°C
Sucrose is widely utilized in medications to carry a more pleasant taste to indigestible chemicals. It is used in many medical dosage forms such as chewable tablets, syrups, tablets, or glues. Sucrose is mainly used in foods nowadays. The fatty acid esters of sucrose show useful application in transdermal therapeutic systems. Sucrose-gradient centrifugation also plays an important role in the selective isolation of nocardia spp. from soil.