Hydriodic acid, ACS, 55-58%, Thermo Scientific™
Hydriodic acid, ACS, 55-58%, Thermo Scientific™
Hydriodic acid, ACS, 55-58%, Thermo Scientific™
Hydriodic acid, ACS, 55-58%, Thermo Scientific™
Thermo Scientific Chemicals
Hydriodic acid, ACS, 55-58%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
Catalog number ALF036484.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHydriodic acid
ColorBrown-Red to Yellow
CAS10034-85-2
Density1.701 g/cm3 (14.195 lb./gal)
Health Hazard 2
Hazard Statement
  • H314 Causes severe skin burns and eye damage.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Used as catalyst for conversion of primary alcohol to alkyl halide
  • Suitable for use in methoxyl determinations.