Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
500 mL
1 L
Catalog number ALF036480.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS161671-34-7
IUPAC Name(1R)-3-(tert-butoxy)-3-oxo-1-phenylpropan-1-aminium
Molecular FormulaC13H20NO2
InChI KeyTYYCBAISLMKLMT-LLVKDONJSA-O
SMILESCC(C)(C)OC(=O)C[C@@H]([NH3+])C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO496.0 to 104.0 %
Refractive index1.4960 to 1.4980 (20°C, 589 nm)
Specific optical rotation+16° to +21° (20°C, 589 nm) (c=1, CHCl3)
View more
It is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids.