Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
4 L
Catalog number ALF036480.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS87-91-2
IUPAC Name1,4-diethyl 2,3-dihydroxybutanedioate
Molecular FormulaC8H14O6
InChI KeyYSAVZVORKRDODB-UHFFFAOYNA-N
SMILESCCOC(=O)C(O)C(O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4445-1.4485 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
View more
It is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids.