Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals
CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number ALF036480.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl sulfoxide
Melting Point18.4°C
Name NoteFor analysis ACS
CAS67-68-5
ColorColorless
View more
It is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids.