Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF036462.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS80-05-7
IUPAC Name4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
Molecular FormulaC15H16O2
InChI KeyIISBACLAFKSPIT-UHFFFAOYSA-N
SMILESCC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Flakes or prills
GC>=96.0 %
Infrared spectrumConforms
Melting point155°C to 160°C
Hexamethylenetetramine is used in the manufacturing of phenolic resins liquid preparations and molding compounds. It finds applications in in brake and clutch linings, abrasive products, non-woven textiles and fireproof materials. It is also used as a drug taken by mouth to prevent recurrent urinary tract infections. It is used as a solid fuel in association with 1,3,5-trioxane in campers, hobbyists, the military and relief organizations. It acts as a food additive and as a preservative. It is an important reagent in organic synthesis, especially Duff reaction, Sommelet reaction and in the Delepine reaction. Further, it is used in adhesives, coatings, sealants, and leather preservatives. It is utilized as a formaldehyde releaser for photographic film development and as a hardener for epoxy resins of the bisphenol A type.