Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF036462.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHexamethylenetetramine
CAS100-97-0
Health Hazard 1H228-H317-H500
Health Hazard 2GHS H Statement
H228-H317
Flammable solid.
May cause an allergic skin reaction.
Health Hazard 3P210-P240-P241-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c
View more
Hexamethylenetetramine is used in the manufacturing of phenolic resins liquid preparations and molding compounds. It finds applications in in brake and clutch linings, abrasive products, non-woven textiles and fireproof materials. It is also used as a drug taken by mouth to prevent recurrent urinary tract infections. It is used as a solid fuel in association with 1,3,5-trioxane in campers, hobbyists, the military and relief organizations. It acts as a food additive and as a preservative. It is an important reagent in organic synthesis, especially Duff reaction, Sommelet reaction and in the Delepine reaction. Further, it is used in adhesives, coatings, sealants, and leather preservatives. It is utilized as a formaldehyde releaser for photographic film development and as a hardener for epoxy resins of the bisphenol A type.