Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
CAS: 57-13-6 | CH4N2O | 60.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number ALF036429.A4
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.75 % (Excluding stabilizer)
UVat 255 nm A: =<0.25
UVat 290 nm A: =<0.01
Residue after evaporation=<0.001 %
View more
Urea has important use as a fertilizer and feed supplement. Used for the preparation of plastics and drugs. Urea and its derivatives plays an important role in the cosmetics industry.