Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
CAS: 57-13-6 | CH4N2O | 60.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number ALF036429.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS91-15-6
IUPAC Namebenzene-1,2-dicarbonitrile
Molecular FormulaC8H4N2
InChI KeyXQZYPMVTSDWCCE-UHFFFAOYSA-N
SMILESN#CC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point138°C to 142°C
GC>=97.5 %
Appearance (Form)Powder or crystals
Appearance (Color)White to beige to grey-green
Infrared spectrumConforms
Urea has important use as a fertilizer and feed supplement. Used for the preparation of plastics and drugs. Urea and its derivatives plays an important role in the cosmetics industry.