Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
CAS: 57-13-6 | CH4N2O | 60.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number ALF036429.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS122-00-9
IUPAC Name1-(4-methylphenyl)ethan-1-one
Molecular FormulaC9H10O
InChI KeyGNKZMNRKLCTJAY-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Appearance (Color)Clear colorless to yellow
Refractive index1.5300 to 1.5350 (20°C, 589 nm)
Urea has important use as a fertilizer and feed supplement. Used for the preparation of plastics and drugs. Urea and its derivatives plays an important role in the cosmetics industry.