Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Used as a protein denaturant
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number ALF036428.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Chemical Identifiers
CAS1321-74-0
IUPAC Name1,2-diethenylbenzene
Molecular FormulaC10H10
InChI KeyMYRTYDVEIRVNKP-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
FormLiquid
Color (APHA)≤500
Assay (unspecified)79.0 to 81.0% (mixture of isomers, meta and para)
Stabilizer900-1100ppm 4-tert-butylcatechol
Urea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.