Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals
Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals
Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals
Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals
CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF036415.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7758-94-3
IUPAC Nameλ²-iron(2+) dichloride
Molecular FormulaCl2Fe
InChI KeyNMCUIPGRVMDVDB-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Fe++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
FormPowder
Total Metal Impurities≤0.5% (≤5000 ppm)
It is used most commonly by athletes to describe the intense pain felt during exhaustive exercise. Used to make cultured dairy products, as a food preservative, and to make chemicals. Useful for the creation of highly aligned electrospun poly-L-lactic acid fibers for nerve regeneration applications. Poly(lactic acid) nanocomposites with various organoclays shows useful application for gas permeability.