Tannic acid, ACS, Thermo Scientific Chemicals
Tannic acid, ACS, Thermo Scientific Chemicals
Tannic acid, ACS, Thermo Scientific Chemicals
Tannic acid, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tannic acid, ACS, Thermo Scientific Chemicals
CAS: 1401-55-4 | C76H52O46 | 1701.206 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF036410.22
Price (MYR)
328.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
328.00
Add to cart
Specifications
Chemical Name or MaterialTannic Acid
CAS1401-55-4
Melting Point∽218°C (decomposition)
Recommended StorageAmbient temperatures
RTECS NumberWW5075000
View more
Tannic acid has anti-bacterial, anti-enzymatic and astringent applications. Used in the dyeing process for cellulose fibers such as cotton. Used in food and medication.