D-Sorbitol, 98%
D-Sorbitol, 98%
D-Sorbitol, 98%
Thermo Scientific Chemicals
D-Sorbitol, 98%
CAS: 50-70-4 | C6H14O6 | 182.172 g/mol
Have Questions?
Catalog NumberQuantity
ALF036404.36500 g
Catalog number ALF036404.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS2039-82-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow
Solution Test(10% v/v in methanol) : Clear
Assay (GC)>97.5%
Solution Test(10% v/v in diethyl ether) : Clear
Refractive Index1.5915-1.5965 @ 20?C
View more
D-sorbitol is added to food products as sweetener. It is also used in industrial and medical applications. Sorbitol often is applied in modern cosmetics as a humectant and thickener.