Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals
CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
Have Questions?
Catalog NumberQuantity
ALF036401.K21 L
Catalog number ALF036401.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS25155-30-0
IUPAC Namesodium 2-dodecylbenzene-1-sulfonate
Molecular FormulaC18H29NaO3S
InChI KeyHFQQZARZPUDIFP-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Assay>=78 %
Water=<2 %
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.