Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 L
100 mL
Catalog number ALF036289.K3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 L
Request bulk or custom format
Chemical Identifiers
CAS98330-04-2
IUPAC Nametetraethylazanium fluoride
Molecular FormulaC8H20FN
InChI KeyQSUJAUYJBJRLKV-UHFFFAOYSA-M
SMILES[F-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)< 28% (non-U.S. specification)
Assay (Non-aqueous acid-base Titration)≥96.0% (dry wt. basis, non-U.S. specification)
FormPowder or crystals or crystalline powder
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥96.0% (U.S. specification)
It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.