Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2 L
Catalog number ALF036289.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic Acid, glacial
Name NoteGlacial
CAS64-19-7
Melting Point16.6°C
Boiling Point118.1°C
View more
It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.