Straight Wall Nickel Crucible
Straight Wall Nickel Crucible
Straight Wall Nickel Crucible
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
250 mL
20 mL
500 mL
Diameter (Metric) Outer:
View all
Outside: 82mm
Catalog number ALF036098.KT
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Diameter (Metric) Outer:
Outside: 82mm
Request bulk or custom format
Specifications
DescriptionStraight Wall
MaterialNickel
Capacity (Metric)250 mL
Diameter (Metric) OuterOutside: 82mm
Height (Metric)45mm