Hydrochloric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035646.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS547-58-0
IUPAC Name4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Molecular FormulaC14H14N3O3S
InChI KeyIETWCRRCPURZOC-UHFFFAOYSA-M
SMILESCN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Visual transition intervalfrom pH 3.2 (pink or red)to pH 4.4 (yellow-orange)
Infrared spectrumConforms
Appearance (Color)Orange
Solubility(0.1 % in water) Clear
View more
Hydrochloric acid, 0.05N standardized solution is used for volumetric analysis in laboratories.