Lactic acid, 1.0N Standardized Solution
Lactic acid, 1.0N Standardized Solution
Lactic acid, 1.0N Standardized Solution
Lactic acid, 1.0N Standardized Solution
Thermo Scientific Chemicals
Lactic acid, 1.0N Standardized Solution
CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF035628.AP500 mL
Catalog number ALF035628.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS119-26-6
IUPAC Name(2,4-dinitrophenyl)hydrazine
Molecular FormulaC6H6N4O4
InChI KeyHORQAOAYAYGIBM-UHFFFAOYSA-N
SMILESNNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
Water30 to 45 % (K.F.)
Appearance (Form)Moist powder
HPLC>=98.0 %