Sodium carbonate, 0.05N Standardized Solution
Sodium carbonate, 0.05N Standardized Solution
Sodium carbonate, 0.05N Standardized Solution
Sodium carbonate, 0.05N Standardized Solution
Thermo Scientific Chemicals
Sodium carbonate, 0.05N Standardized Solution
CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Catalog NumberQuantity
ALF035609.K21 L
Catalog number ALF035609.K2
Price (MYR)
333.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
333.00
Add to cart
Specifications
Chemical Name or MaterialSodium carbonate
CAS497-19-8
Recommended StorageAmbient temperatures
Physical FormLiquid
Quantity1 L