D-Mannitol, ACS
D-Mannitol, ACS
D-Mannitol, ACS
Thermo Scientific Chemicals
D-Mannitol, ACS
CAS: 69-65-8 | C6H14O6 | 182.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Catalog number ALF033342.A4
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS111-78-4
IUPAC Name(1Z,5Z)-cycloocta-1,5-diene
Molecular FormulaC8H12
InChI KeyVYXHVRARDIDEHS-QGTKBVGQSA-N
SMILESC1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4920 to 1.4950 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Stabilizer100 to 200 ppm Octadecyl 3-(3',5'-di-tert-butyl
Stabilizer-4'-hydroxyphenyl)propionate
View more
D-Mannitol is used with boric acid to make dry electrolytic condensers for radio applications, artificial resins and plasticizers. It is used as excipient for chewable tablets due to its pleasant taste. It is used in volumetric analysis of boric acid in analytical chemistry. It is also used in the food industry as anticaking, free-flow agent, flavoring agent, stabilizer, thickener and nutritive sweetener. Further, it is used in medicine as diuretic and in the determination of renal function. In addition, it is used as a formulation aid, lubricant, non-nutritive sweetener and solvent for foods.