Dithizone, ACS, 85+%, Thermo Scientific Chemicals
Dithizone, ACS, 85+%, Thermo Scientific Chemicals
Dithizone, ACS, 85+%, Thermo Scientific Chemicals
Dithizone, ACS, 85+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dithizone, ACS, 85+%, Thermo Scientific Chemicals
CAS: 60-10-6 | C13H12N4S | 256.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number ALF033283.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDithizone
CAS60-10-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Dithizone is used as an indicator for minute amounts of lead. It acts as a chelating agent for heavy metal poisoning. As a ligand, it forms complexes with metals like zinc, lead and mercury. Further, it is used to assess the purity of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes. In addition to this, it is used as a titrant in the quantitative determination of heavy metals.