Sulfuric acid, ACS, 95.0-98.0%, Thermo Scientific Chemicals
Sulfuric acid, ACS, 95.0-98.0%, Thermo Scientific Chemicals
Sulfuric acid, ACS, 95.0-98.0%, Thermo Scientific Chemicals
Sulfuric acid, ACS, 95.0-98.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, ACS, 95.0-98.0%, Thermo Scientific Chemicals
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
4 x 1 kg
Catalog number ALF033273.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS16423-68-0
IUPAC Namedisodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6I4Na2O5
InChI KeyIINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Dye content >= 87.0%
Solution Test1% in water: clear
Appearance (Color)Red to red-brown
lambda-max (UV-Vis)524-528nm (water)
FormPowder
Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.