Silicic acid hydrate, Thermo Scientific Chemicals
Silicic acid hydrate, Thermo Scientific Chemicals
Silicic acid hydrate, Thermo Scientific Chemicals
Silicic acid hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silicic acid hydrate, Thermo Scientific Chemicals
CAS: 1343-98-2 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF033270.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS2627-95-4
IUPAC Nameethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane
Molecular FormulaC8H18OSi2
InChI KeyBITPLIXHRASDQB-UHFFFAOYSA-N
SMILESC[Si](C)(O[Si](C)(C)C=C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4100 to 1.4130 (20°C, 589 nm)
Appearance (Color)Clear colorless
Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.