Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™
Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™
Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™
Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™
Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™
Thermo Scientific Chemicals
Phosphoric acid, 85% w/w aq. soln., ACS, Thermo Scientific™
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Catalog NumberQuantity
ALF033266.A11 kg
Catalog number ALF033266.A1
Price (MYR)
443.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
443.00
Add to cart
Specifications
Chemical Name or MaterialPhosphoric acid
Melting Point19°C to 21°C
CAS7664-38-2
ColorColorless
Health Hazard 1H290-H314
View more
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and in leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion into a water soluble phosphate compound.